5-Methyl-1-(propan-2-yl)-1H-pyrazole-3-carboxylic acid

Leverandør: Apollo Scientific
Warning

OR306338-250MG
APOSOR306338-250MGEA 6275 NOK
APOSOR306338-250MG
5-Methyl-1-(propan-2-yl)-1H-pyrazole-3-carboxylic acid
5-Methyl-1-(propan-2-yl)-1H-pyrazole-3-carboxylic acid
Formel: C₈H₁₂N₂O₂
MW: 168,193 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD08700675
CAS-nummer: 50920-49-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR