Boc-D-α-methylserine

Leverandør: Apollo Scientific
Warning

OR307056-1G
APOSOR307056-1GEA 7800 NOK
APOSOR307056-1G
Boc-D-α-methylserine
Boc-D-alpha-methylserine
Formel: C₉H₁₇NO₅
MW: 219.24 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02682596
CAS-nummer: 84311-18-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR