N-Boc-cis-4-hydroxy-L-proline 95+%

Leverandør: Apollo Scientific
Warning

OR307072-100G OR307072-25G OR307072-5G
APOSOR307072-100GEA 2200 NOK
APOSOR307072-100G APOSOR307072-25G APOSOR307072-5G
N-Boc-cis-4-hydroxy-L-proline 95+%
N-Boc-cis-4-hydroxy-L-proline
Formel: C₁₀H₁₇NO₅
Smeltepunkt: 146 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02094406
CAS-nummer: 87691-27-8
FN: 3077
ADR: 9+EHS,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR