cis-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected 95+%

Leverandør: Apollo Scientific
Warning

Synonymer: tert-Butyl (cis-3-carboxycyclopent-1-yl)carbamate, cis-3-(Boc-amino)cyclopentanecarboxylic acid, cis-3-[(tert-Butoxycarbonyl)amino]cyclopentane-1-carboxylic acid

OR307106-1G
APOSOR307106-1GEA 1050 NOK
APOSOR307106-1G
cis-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected 95+%
cis-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected
Formel: C₁₁H₁₉NO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09992879
CAS-nummer: 410090-37-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR