(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid

Leverandør: Apollo Scientific

Synonymer: (1R,2R)-Cyclohexane-1,2-dicarboxylic acid, (1R,2R)-(-)-trans-Cyclohexane-1,2-dicarboxylic acid

OR307140-100G
APOSOR307140-100GEA 1475 NOK
APOSOR307140-100G
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid
Formel: C₈H₁₂O₄
MW: 172.18 g/mol
Smeltepunkt: 184 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00062973
CAS-nummer: 46022-05-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR