cis-2-Amino-cyclopentanecarboxylic acid hydrochloride

Leverandør: Apollo Scientific

OR307527-1G
APOSOR307527-1GEA 6925 NOK
APOSOR307527-1G
cis-2-Amino-cyclopentanecarboxylic acid hydrochloride
cis-2-Amino-cyclopentanecarboxylic acid hydrochloride
Formel: C₆H₁₁NO₂·ClH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05863555
CAS-nummer: 18414-30-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR