3-Aza-tricyclo[4.2.1.0*2,5*]non-7-en-4-one

Leverandør: Apollo Scientific

Synonymer: 4-Oxo-3-aza-tricyclo[4.2.1.0(2.5)]non-7-ene

OR307552-1G
APOSOR307552-1GEA 5750 NOK
APOSOR307552-1G
3-Aza-tricyclo[4.2.1.0*2,5*]non-7-en-4-one
3-Aza-tricyclo[4.2.1.0*2,5*]non-7-en-4-one
Formel: C₈H₉NO
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00143363
CAS-nummer: 14735-70-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR