cis-2-tert-Butoxycarbonylamino-2-methyl-cyclopentanecarboxylic acid

Leverandør: Apollo Scientific
Warning

OR307584-1G OR307584-250MG
APOSOR307584-1GEA 7975 NOK
APOSOR307584-1G APOSOR307584-250MG
cis-2-tert-Butoxycarbonylamino-2-methyl-cyclopentanecarboxylic acid
cis-2-tert-Butoxycarbonylamino-2-methyl-cyclopentanecarboxylic acid
Formel: C₁₂H₂₁NO₄
Smeltepunkt: 101…102 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05863640
CAS-nummer: 1212127-10-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR