cis-2-Amino-cyclooctanecarboxylic acid

Leverandør: Apollo Scientific

OR307648-1G
APOSOR307648-1GEA 6125 NOK
APOSOR307648-1G
cis-2-Amino-cyclooctanecarboxylic acid
cis-2-Amino-cyclooctanecarboxylic acid
Formel: C₉H₁₇NO₂
MW: 171.237 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD08437545
CAS-nummer: 649765-17-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR