cis-2-tert-Butoxycarbonylamino-cyclooctanecarboxylic acid

Leverandør: Apollo Scientific

OR307658-250MG
APOSOR307658-250MGEA 3600 NOK
APOSOR307658-250MG
cis-2-tert-Butoxycarbonylamino-cyclooctanecarboxylic acid
cis-2-tert-Butoxycarbonylamino-cyclooctanecarboxylic acid
Formel: C₁₄H₂₅NO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09475429
CAS-nummer: 1013980-15-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR