7-Chloro-2-methyl-quinoline-3-carboxylic acid ethyl ester

Leverandør: Apollo Scientific

OR307812-1G OR307812-5G
APOSOR307812-1GEA 2900 NOK
APOSOR307812-1G APOSOR307812-5G
7-Chloro-2-methyl-quinoline-3-carboxylic acid ethyl ester
7-Chloro-2-methyl-quinoline-3-carboxylic acid ethyl ester
Formel: C₁₃H₁₂ClNO₂
MW: 249.693 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05863550
CAS-nummer: 733719-74-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR