7-Chloroquinoline-3-carboxylic acid

Leverandør: Apollo Scientific

OR307836-1G
APOSOR307836-1GEA 6625 NOK
APOSOR307836-1G
7-Chloroquinoline-3-carboxylic acid
7-Chloroquinoline-3-carboxylic acid
Formel: C₁₀H₆ClNO₂
MW: 207,613 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD08437567
CAS-nummer: 892874-49-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR