2-Boc-6-Amino-2-aza-spiro[3.3]heptane

Leverandør: Apollo Scientific
Warning

Synonymer: 6-Amino-2-azaspiro[3.3]heptane, N2-BOC protected, tert-Butyl 6-amino-2-azaspiro[3.3]heptane-2-carboxylate

OR308106-100MG
APOSOR308106-100MGEA 680 NOK
APOSOR308106-100MG
2-Boc-6-Amino-2-aza-spiro[3.3]heptane
2-Boc-6-amino-2-azaspiro[3.3]heptane
Formel: C₁₁H₂₀N₂O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD15071432
CAS-nummer: 1211586-09-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR