2-Benzylmorpholine

Leverandør: Apollo Scientific

OR308189-1G
APOSOR308189-1GEA 4175 NOK
APOSOR308189-1G
2-Benzylmorpholine
2-Benzylmorpholine
Formel: C₁₁H₁₅NO
MW: 177,243 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09055007
CAS-nummer: 87955-28-0

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR