(2R)-2-Benzylmorpholine

Leverandør: Apollo Scientific

Synonymer: (R)-2-Benzylmorpholine

OR308191-250MG
APOSOR308191-250MGEA 1850 NOK
APOSOR308191-250MG
(2R)-2-Benzylmorpholine
(2R)-2-Benzylmorpholine
Formel: C₁₁H₁₅NO
MW: 177,243 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD14635495
CAS-nummer: 131887-51-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR