3-(3-Bromophenyl)propionic acid 97%

Leverandør: Apollo Scientific

OR3084-100G
APOSOR3084-100GEA 2000 NOK
APOSOR3084-100G
3-(3-Bromophenyl)propionic acid 97%
3-(3-Bromophenyl)propionic acid
Formel: C₉H₉BrO₂
MW: 229.07 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01310792
CAS-nummer: 42287-90-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR