3-(4-Bromophenyl)propionic acid

Leverandør: Apollo Scientific

OR3085-100G
APOSOR3085-100GEA 1500 NOK
APOSOR3085-100G
3-(4-Bromophenyl)propionic acid
3-(4-Bromophenyl)propionic acid
Formel: C₉H₉BrO₂
MW: 229.07 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01310793
CAS-nummer: 1643-30-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR