7-Bromo-2-hydroxy-3-phenylquinoline

Leverandør: Apollo Scientific

OR309276-1G
APOSOR309276-1GEA 6125 NOK
APOSOR309276-1G
7-Bromo-2-hydroxy-3-phenylquinoline
7-Bromo-2-hydroxy-3-phenylquinoline
Formel: C₁₅H₁₀BrNO
MW: 300,15 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09997966
CAS-nummer: 861872-50-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR