7-Bromo-4-hydroxy-2-phenylquinoline

Leverandør: Apollo Scientific
Danger

OR309302-1G
APOSOR309302-1GEA 4925 NOK
APOSOR309302-1G
7-Bromo-4-hydroxy-2-phenylquinoline
7-Bromo-4-hydroxy-2-phenylquinoline
Formel: C₁₅H₁₀BrNO
MW: 300,15 g/mol
Smeltepunkt: 280 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11505233
CAS-nummer: 825620-24-4
FN: 2811
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR