5-Chloro-3-[(3,4-dimethoxyphenyl)methyl]-1,2,4-thiadiazole

Leverandør: Apollo Scientific

OR309415-1G
APOSOR309415-1GEA 6675 NOK
APOSOR309415-1G
5-Chloro-3-[(3,4-dimethoxyphenyl)methyl]-1,2,4-thiadiazole
5-Chloro-3-[(3,4-dimethoxyphenyl)methyl]-1,2,4-thiadiazole
Formel: C₁₁H₁₁ClN₂O₂S
MW: 270,735 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD14707328
CAS-nummer: 1221342-59-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR