cis-2-Amino-2-methyl-cyclopentanecarboxylic acid hydrochloride

Leverandør: Apollo Scientific

OR309428-1G
APOSOR309428-1GEA 3900 NOK
APOSOR309428-1G
cis-2-Amino-2-methyl-cyclopentanecarboxylic acid hydrochloride
cis-2-Amino-2-methyl-cyclopentanecarboxylic acid hydrochloride
Formel: C₇H₁₃NO₂·ClH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05863639
CAS-nummer: 156292-34-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR