cis-1-Amino-1,2,3,4-tetrahydro-2-naphthalenecarboxylic acid

Leverandør: Apollo Scientific

OR309433-250MG
APOSOR309433-250MGEA 4125 NOK
APOSOR309433-250MG
cis-1-Amino-1,2,3,4-tetrahydro-2-naphthalenecarboxylic acid
cis-1-Amino-1,2,3,4-tetrahydro-2-naphthalenecarboxylic acid
Formel: C₁₁H₁₃NO₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09260408
CAS-nummer: 888407-44-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR