3,4-Dibromo-1-methyl-2,5-dihydro-1H-pyrrole-2,5-dione

Leverandør: Apollo Scientific
Warning

Synonymer: 2,3-Dibromo-N-methylmaleimide

OR310340-1G
APOSOR310340-1GEA 2175 NOK
APOSOR310340-1G
3,4-Dibromo-1-methyl-2,5-dihydro-1H-pyrrole-2,5-dione
3,4-Dibromo-1-methyl-2,5-dihydro-1H-pyrrole-2,5-dione
Formel: C₅H₃Br₂NO₂
MW: 268,891 g/mol
Smeltepunkt: 120…124 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00102284
CAS-nummer: 3005-27-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR