2-Methylpropane-1,2-diol

Leverandør: Apollo Scientific
Warning

Synonymer: Dimethyl glyoxal

OR310412-1G
APOSOR310412-1GEA 270 NOK
APOSOR310412-1G
2-Methylpropane-1,2-diol
2-Methylpropane-1,2-diol
Formel: C₄H₁₀O₂
MW: 90.121 g/mol
Kokepunkt: 177…178 °C
Tetthet: 0.997 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00971926
CAS-nummer: 558-43-0
EINECS: 209-198-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR