3-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzaldehyde

Leverandør: Apollo Scientific

OR310499-5G
APOSOR310499-5GEA 2950 NOK
APOSOR310499-5G
3-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzaldehyde
3-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzaldehyde
Formel: C₁₂H₁₂BNO₅
MW: 261,038 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD11215223
CAS-nummer: 1488424-86-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR