1-Pentyl-1H-indole-3-carboxylic acid

Leverandør: Apollo Scientific

OR310721-1G
APOSOR310721-1GEA 9600 NOK
APOSOR310721-1G
1-Pentyl-1H-indole-3-carboxylic acid
1-Pentyl-1H-indole-3-carboxylic acid
Formel: C₁₄H₁₇NO₂
MW: 231.3 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD05859192
CAS-nummer: 727421-73-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR