4-Bromophenylboronic acid neopentyl glycol ester

Leverandør: Apollo Scientific

Synonymer: 2-(4-Bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane, 4-Bromobenzeneboronic acid neopentyl glycol ester

OR310880-5G
APOSOR310880-5GEA 1150 NOK
APOSOR310880-5G
4-Bromophenylboronic acid neopentyl glycol ester
4-Bromophenylboronic acid neopentyl glycol ester
Formel: C₁₁H₁₄BBrO₂
MW: 268.94 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01318105
CAS-nummer: 183677-71-6

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR