4-{[3-(4-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]methyl}piperidine hydrochloride

Leverandør: Apollo Scientific

OR310994-1G
APOSOR310994-1GEA 9600 NOK
APOSOR310994-1G
4-{[3-(4-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]methyl}piperidine hydrochloride
4-{[3-(4-Methoxyphenyl)-1,2,4-oxadiazol-5-yl]methyl}piperidine hydrochloride
Formel: C₁₅H₁₉N₃O₂·ClH
MW: 309.791 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD28041319
CAS-nummer: 1622069-60-6

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR