2,2'-Bipyrimidyl

Leverandør: Apollo Scientific
Warning

Synonymer: 2,2'-Dipyrimidyl, 2,2'-Bipyrimidine

OR315153-1G
APOSOR315153-1GEA 680 NOK
APOSOR315153-1G
2,2'-Bipyrimidyl
2,2'-Bipyrimidyl
Formel: C₈H₆N₄
MW: 158.16 g/mol
Smeltepunkt: 112…116 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00014600
CAS-nummer: 34671-83-5
EINECS: 252-137-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR