(4-Dimethylaminophenyl)di-tert-butylphosphine 97%

Leverandør: Apollo Scientific
Warning

Synonymer: 4-(Di-tert-butylphosphino)-N,N-dimethylaniline

OR315484-5G
APOSOR315484-5GEA 380 NOK
APOSOR315484-5G
(4-Dimethylaminophenyl)di-tert-butylphosphine 97%
(4-Dimethylaminophenyl)di-tert-butylphosphine
Formel: C₁₆H₂₈NP
MW: 265,38 g/mol
Smeltepunkt: 57…61 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09265102
CAS-nummer: 932710-63-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR