1,2-Bis(difenylfosfino)etan

Leverandør: Apollo Scientific
Warning

Synonymer: Ethylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)ethane, DIPHOS, 1,2-Bis(diphenylphosphino)ethane, Ethane-1,3-diylbis(diphenylphosphine)

OR315704-100G OR315704-25G
APOSOR315704-100GEA 415 NOK
APOSOR315704-100G APOSOR315704-25G
1,2-Bis(difenylfosfino)etan
1,2-Bis(difenylfosfino)etan
Formel: C₂₆H₂₄P₂
MW: 398.42 g/mol
Smeltepunkt: 137…142 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003047
CAS-nummer: 1663-45-2
EINECS: 216-769-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR