1,5-Bis(difenylfosfino)pentan 97%

Leverandør: Apollo Scientific

Synonymer: Pentane-1,3-diylbis(diphenylphosphine), Pentylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)pentane

OR315706-25G OR315706-10G
APOSOR315706-25GEA 0 NOK
APOSOR315706-25G APOSOR315706-10G
1,5-Bis(difenylfosfino)pentan 97%
1,5-Bis(difenylfosfino)pentan
Formel: C₂₉H₃₀P₂
MW: 440.5 g/mol
Kokepunkt: 230
Smeltepunkt: 42…44 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003052
CAS-nummer: 27721-02-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR