7-Aza-spiro[3.5]nonane hydrochloride

Leverandør: Apollo Scientific

Synonymer: 7-Azaspiro[3.5]nonane hydrochloride

OR317147-1G
APOSOR317147-1GEA 4100 NOK
APOSOR317147-1G
7-Aza-spiro[3.5]nonane hydrochloride
7-Aza-spiro[3.5]nonane hydrochloride
Formel: C₈H₁₅N·ClH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD23105809
CAS-nummer: 1414885-16-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR