3-Boc-6-oxo-3-aza-bicyclo[3.1.1]heptane

Leverandør: Apollo Scientific

OR317281-100MG OR317281-250MG
APOSOR317281-100MGEA 1550 NOK
APOSOR317281-100MG APOSOR317281-250MG
3-Boc-6-oxo-3-aza-bicyclo[3.1.1]heptane
3-Boc-6-oxo-3-aza-bicyclo[3.1.1]heptane
Formel: C₁₁H₁₇NO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD17016750
CAS-nummer: 1251013-26-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR