4-tert-Butoxybenzoic acid 95+%

Leverandør: Apollo Scientific

Synonymer: 4-(tert-Butoxy)benzoic acid

OR318017-1G OR318017-25G OR318017-5G
APOSOR318017-1GEA 1100 NOK
APOSOR318017-1G APOSOR318017-25G APOSOR318017-5G
4-tert-Butoxybenzoic acid 95+%
4-tert-Butoxybenzoic acid
Formel: C₁₁H₁₄O₃
MW: 194.23 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00082602
CAS-nummer: 13205-47-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR