2,6,6-Trimethyl-4,5,6,7-tetrahydro-1-benzofuran-4-one 95%

Leverandør: Apollo Scientific

OR32030-1G
APOSOR32030-1GEA 19275 NOK
APOSOR32030-1G
2,6,6-Trimethyl-4,5,6,7-tetrahydro-1-benzofuran-4-one 95%
2,6,6-Trimethyl-4,5,6,7-tetrahydro-1-benzofuran-4-one
Formel: C₁₁H₁₄O₂
MW: 178,228 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00215387
CAS-nummer: 57822-05-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR