N'-[(1E)-1-(4-Chlorophenyl)ethylidene]methoxycarbohydrazide, tech

Leverandør: Apollo Scientific

OR32069-1G
APOSOR32069-1GEA 19275 NOK
APOSOR32069-1G
N'-[(1E)-1-(4-Chlorophenyl)ethylidene]methoxycarbohydrazide, tech
N'-[(1E)-1-(4-Chlorophenyl)ethylidene]methoxycarbohydrazide
Formel: C₁₀H₁₁ClN₂O₂
MW: 226,66 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00792941
CAS-nummer: 95855-11-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR