2-{[(4-Bromophenyl)amino]methylidene}cyclohexane-1,3-dione, tech

Leverandør: Apollo Scientific

OR32097-1G
APOSOR32097-1GEA 19275 NOK
APOSOR32097-1G
2-{[(4-Bromophenyl)amino]methylidene}cyclohexane-1,3-dione, tech
2-{[(4-Bromophenyl)amino]methylidene}cyclohexane-1,3-dione
Formel: C₁₃H₁₂BrNO₂
MW: 294,144 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01569054
CAS-nummer: 341968-26-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR