(Z)-Ethyl-3-(pyrrolidin-1-yl)but-2-enoate 95%

Leverandør: Apollo Scientific

Synonymer: (Z)-Ethyl 3-(pyrrolidin-1-yl)but-2-enoate

OR32104-1G
APOSOR32104-1GEA 1900 NOK
APOSOR32104-1G
(Z)-Ethyl-3-(pyrrolidin-1-yl)but-2-enoate 95%
(Z)-Ethyl-3-(pyrrolidin-1-yl)but-2-enoate
Formel: C₁₀H₁₇NO₂
MW: 183,25 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02081922
CAS-nummer: 70526-06-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR