(R)-4,5-Dimethyl-2-(pyrrolidin-2-yl)thiazole hydrobromide 95%

Leverandør: Apollo Scientific
Warning

OR321281-100MG
APOSOR321281-100MGEA 1700 NOK
APOSOR321281-100MG
(R)-4,5-Dimethyl-2-(pyrrolidin-2-yl)thiazole hydrobromide 95%
(R)-4,5-Dimethyl-2-(pyrrolidin-2-yl)thiazole hydrobromide
Formel: C₉H₁₄N₂S·BrH
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD30728434
CAS-nummer: 1821837-31-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR