rac-(R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-3-yl)acetic acid

Leverandør: Apollo Scientific

OR321538-1G OR321538-250MG
APOSOR321538-1GEA 4925 NOK
APOSOR321538-1G APOSOR321538-250MG
rac-(R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-3-yl)acetic acid
rac-(R)-2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)pyrrolidin-3-yl)acetic acid
Formel: C₂₁H₂₁NO₄
MW: 351.396 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD02179121
CAS-nummer: 868523-86-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR