(2E)-3-(Dimethylamino)-1-(5-methylfuran-2-yl)prop-2-en-1-one 95%

Leverandør: Apollo Scientific

OR32192-1G
APOSOR32192-1GEA 10750 NOK
APOSOR32192-1G
(2E)-3-(Dimethylamino)-1-(5-methylfuran-2-yl)prop-2-en-1-one 95%
(2E)-3-(Dimethylamino)-1-(5-methylfuran-2-yl)prop-2-en-1-one
Formel: C₁₀H₁₃NO₂
MW: 179,216 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00172111
CAS-nummer: 1233858-29-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR