4,6-Dimethyl-2-hydroxypyrimidine 97%

Leverandør: Apollo Scientific

Synonymer: 2-Hydroxy-4,6-dimethylpyrimidine, 4,6-Dimethyl-2-pyrimidinol, 4,6-Dimethylpyrimidin-2(1H)-one

OR322007-100G
APOSOR322007-100GEA 805 NOK
APOSOR322007-100G
4,6-Dimethyl-2-hydroxypyrimidine 97%
4,6-Dimethyl-2-hydroxypyrimidine
Formel: C₆H₈N₂O
MW: 124.14 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006072
CAS-nummer: 108-79-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR