2,2,5,7,8-Pentamethyl-6-chromanol

Leverandør: Apollo Scientific

Synonymer: 2,2,5,7,8-Pentamethylchroman-6-ol

OR322018-1G
APOSOR322018-1GEA 520 NOK
APOSOR322018-1G
2,2,5,7,8-Pentamethyl-6-chromanol
2,2,5,7,8-Pentamethyl-6-chromanol
Formel: C₁₄H₂₀O₂
MW: 220,307 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00210347
CAS-nummer: 950-99-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR