Methyl-2,2-dimethylacetoacetate

Leverandør: Apollo Scientific

Synonymer: Methyl 2,2-dimethylacetoacetate

OR322264-1G
APOSOR322264-1GEA 790 NOK
APOSOR322264-1G
Methyl-2,2-dimethylacetoacetate
Methyl-2,2-dimethylacetoacetate
Formel: C₇H₁₂O₃
MW: 144.17 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00026232
CAS-nummer: 38923-57-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR