Dietylentriamin

Leverandør: Apollo Scientific

Synonymer: 2,2'-Iminodi(ethylamine), DIEN, Diethylenetriamine

OR322494-100G
APOSOR322494-100GEA 0 NOK
APOSOR322494-100G
Dietylentriamin
Dietylentriamin
Formel: C₄H₁₃N₃
MW: 103,17 g/mol
Kokepunkt: 206 °C
Smeltepunkt: –40 °C
Tetthet: 0,96 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008171
CAS-nummer: 111-40-0
FN: 2079
ADR: 8,II

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR