N-Succinimidyl 4-Formylbenzoate

Leverandør: Apollo Scientific

Synonymer: Ald-Ph-NHS ester, 2,5-Dioxopyrrolidin-1-yl 4-formylbenzoate, 4-Formylbenzoic Acid N-Succinimidyl Ester

OR323071-1G
APOSOR323071-1GEA 0 NOK
APOSOR323071-1G
N-Succinimidyl 4-Formylbenzoate
N-Succinimidyl 4-Formylbenzoate
Formel: C₁₂H₉NO₅
MW: 247.21 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00133733
CAS-nummer: 60444-78-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR