1-Amino-3-phenylpropan-2-ol 95%

Leverandør: Apollo Scientific

OR323295-250MG
APOSOR323295-250MGEA 1200 NOK
APOSOR323295-250MG
1-Amino-3-phenylpropan-2-ol 95%
1-Amino-3-phenylpropan-2-ol
Formel: C₉H₁₃NO
MW: 151,206 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03840182
CAS-nummer: 50411-26-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR