2-(3,4-Dichlorophenyl)-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one, tech

Leverandør: Apollo Scientific

OR32582-1G
APOSOR32582-1GEA 19275 NOK
APOSOR32582-1G
2-(3,4-Dichlorophenyl)-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one, tech
2-(3,4-Dichlorophenyl)-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one
Formel: C₁₂H₁₂Cl₂N₂O₂
MW: 287,142 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03848420
CAS-nummer: 861210-29-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR