2-(2-Chloro-3-pyridinyl)-1H-1,3-benzimidazole 95%

Leverandør: Apollo Scientific

OR32627-1G
APOSOR32627-1GEA 8275 NOK
APOSOR32627-1G
2-(2-Chloro-3-pyridinyl)-1H-1,3-benzimidazole 95%
2-(2-Chloro-3-pyridinyl)-1H-1,3-benzimidazole
Formel: C₁₂H₈ClN₃
MW: 229,67 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD12025828
CAS-nummer: 143426-40-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR